Learn More
Ambu™ Res-Cue Mask Non-distribution product as customer accommodation.
Effective Contact Prevention for mouth-to-mouth resuscitation
Manufacturer:  Ambu™ 000252104
 View more versions of this product
Catalog No. 19-065-695
Add to cart
Description
Description
  • Transparent dome
  • Pocket-size CPR mouth barrier mask
  • Completely seals easily to the face of infants, children, and adults
  • Optional oxygen inlet valve allows for oxygen delivery
  • Reusable mask; easy to clean
  • Replaceable one-way valve/filter in one housing
  • Pliable in extreme temperatures
Specifications
Specifications
Nylon
Reusable mask, one way valve and filter, 1 pair Vinyl gloves, 2 Alcohol wipes, Oxygen inlet and head strap in red soft Plastic case
Each
0.07kg
Rescue mask red soft case
Red
PVC
0.15 lb.
Protected rescue breathing for any patient and may also be used with a resuscitator