Learn More
Ambu™ Res-Cue Mask Non-distribution product as customer accommodation.
Effective Contact Prevention for mouth-to-mouth resuscitation
Manufacturer:  Ambu™ 000252123
 View more versions of this product
Catalog No. 19-065-698
Add to cart
Description
Description
  • Transparent dome
  • Pocket-size CPR mouth barrier mask
  • Completely seals easily to the face of infants, children, and adults
  • Optional oxygen inlet valve allows for oxygen delivery
  • Reusable mask; easy to clean
  • Replaceable one-way valve/filter in one housing
  • Pliable in extreme temperatures
Specifications
Specifications
Nylon
PVC
0.15 lb.
Protected rescue breathing for any patient and may also be used with a resuscitator
Reusable mask, one way valve and filter, 1 pair Vinyl gloves and 2 Alcohol wipes in red soft Plastic case
Each
0.07kg
Rescue mask red soft case