Ambu Res-Cue Mask

Effective Contact Prevention for mouth-to-mouth resuscitation

Manufacturer: Ambu, Inc. 000 252 102

 View more versions of this product

Catalog No. 19-805-925

  Check Availability
Add to Cart

    Specifications

    Case Material Polypropylene
    Color Red
    Includes Reusable mask, one way valve and filter, 1 pair Vinyl gloves, 2 Alcohol wipes, Oxygen inlet and head strap in red hard Plastic case
    Material PVC
    Quantity Each
    Weight (English) 0.26 lb.
    Weight (Metric) 0.11kg
    For Use With (Application) Protected rescue breathing for any patient and may also be used with a resuscitator
    Product Type Rescue mask complete red hard case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit