Learn More
Aat BioquestSupplier Diversity Partner AMCA acid *CAS#: 106562-32-7* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 501
AMCA is one of the brightest blue fluorescent labeling dyes. It can be well excited at 365 nm. It is quite photostable, and its fluorescence is pH-independent from pH 4 to 10.
Catalog No. 50-195-2914