Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ AMD-3+ Multiple Delivery 6psi 3 Flowmeter Inhalant Anesthesia With O2 Concentrator + EPS-3 Exposure Prevention Package
SDP

Gives the user the ability to deliver accurate flowrates, actively scavenge waste gas and the convenience of an Oxygen Concentrator

Supplier:  SOMNI Scientific™ AMD17108

Catalog No. 04-777-106

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

  • 3 Flowmeter Controlled Stations with Toggle Switches
    • Common Gas outlets for each station
    • Adjustable flowrates
    • Three 6 ft. color coded delivery lines with adapters for each station
  • TT Frame
  • SOMNI™ 3 6psi calibrated Isoflurane vaporizer
  • Oxygen Concentrator
    • Quiet Operation
    • Triple Filtration
    • Low cost of ownership
    • Safety alarms
    • Concentrator kit includes single e-cylinder manifold for backup
  • EPS-3 exposure prevention system
    • Actively remove waste gas on multiple devices
    • Energy saving timer, selectable up to 12 hours with auto power off
  • Mobile Workstation
  • Active Induction Chamber 
  • Small footprint
  • Stacking design save valuable workbench space
  • WAG filters
  • UniFlow Nosecone
    • Magnetic placement included
Specifications

Specifications

Research Animal Inhalant Anesthesia
Aluminum, Brass, Stainless Steel, Plastic
110 V
110 V
AMD-3+ 6 psi, TT Frame, SOMNI™ 3 6 psi Iso Vaporizer, Oxygen Concentrator, EPS-3, Workstation, Induction Chamber, WAG Filters, UniFlow Nosecone
EM-16002, EM-16003, EM-16004, EM-16005, EM-16006, EM-16007, EM-16008, EM-16010, EM-16011
53.34 cm
53.34 cm
60 Hz
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Veterinary Use Only