Learn More
Aviva Systems Biology Corp AMDHD1 Antibody - N-terminal region (ARP52962_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMDHD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AMDHD1. Alternate names: HMFT1272, MGC35366ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60880