Amerex™ 5 lb. Multipurpose Extinguishers Available on GSA/VA Contract for Federal Government customers only.

Model B402

Manufacturer: Amerex CorporationSupplier Diversity Partner B500T

 View more versions of this product

Catalog No. 19-029172

  Check Availability
Add to Cart

    Specifications

    Capacity 5 lb., 2.26kg
    Dimensions (L x W x H) 4.25 x 7.25 x 15.25 in. (10.8 x 18.4 x 38.7cm)
    Discharge Time 14 sec.
    Includes Aluminium valve, vehicle/marine mount bracket
    Length (Metric) 10.8cm
    Material Aluminum
    Reagent Type Multipurpose ABC dry chemical
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -53.8° to 48.8°C
    UL Rating 2A:10B:C
    Width (English) 7.25 in.
    Capacity (English) 5 lb.
    Capacity (Metric) 2.26kg
    Product Type Fire extinguisher
    Height (English) 15.25 in.
    Height (Metric) 38.7cm
    Length (English) 4.25 in.
    Model B500T
    Range 12 to 18 ft.
    Width (Metric) 18.4cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit