Amerex™ ABC Dry Chemical Fire Extinguishers

Rugged design with all metal valve construction

Manufacturer: Amerex CorporationSupplier Diversity Partner B441CR

 View more versions of this product

Catalog No. 19-042-350

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Reagent Type ABC dry chemical
    Width (English) 8.75 in.
    Capacity (Metric) 4.5kg
    Capacity 10 lb., 4.5kg
    Dimensions (L x W x H) 5 x 8.75 x 20.5 in. (12.7 x 22.2 x 52cm)
    Discharge Time 20 sec.
    Height (Metric) 52cm
    Includes Wall bracket
    Length (English) 5 in.
    Length (Metric) 12.7cm
    Material Brass, Chrome
    Range 15 to 21 ft.
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -53.8° to 48.8°C
    Capacity (English) 10 lb.
    Height (English) 20.5 in.
    Model B441CR
    UL Rating 4A:80B:C
    Width (Metric) 22.2cm
    Product Type Fire extinguisher

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit