Learn More
Amerex™ ABC ExtinguisherChemical Agent Refill
Refills for Amerex ABC dry chemical fire extinguishers
Manufacturer:  Amerex™ 550
 View more versions of this product
Catalog No. 19-806-003
Add to cart
Description
Description
Refill ABC extinguishers with specially fluidized and siliconized mono ammonium phosphate.
  • Coats surface to which it is applied
  • Will not conduct electricity back to operator
  • Chemically insulates Class A fires by melting at approximately 177°C (350°F)
  • Smothers and breaks chain reaction of Class B fires
Specifications
Specifications
50 lb., 23kg
50 lb.
Dry Monoammonium Phosphate refill
ABC fire extinguisher
23kg
Safety