Amerex™ Halotron I Clean-Agent Stored-Pressure Fire Extinguishers Available on GSA/VA Contract for Federal Government customers only.

Most widely used Halon 1211 replacement agent

Manufacturer: Amerex CorporationSupplier Diversity Partner B385TS

 View more versions of this product

Catalog No. 19-805-991

  Check Availability
Add to Cart

    Specifications

    Capacity 2.5 lb., 1.1kg
    Dimensions (L x W x H) 3 x 6.38 x 15.5 in. (7.62 x 16.2 x 39.3cm)
    Discharge Time 9 sec.
    Includes Anodized Aluminium valve, nozzle, vehicle bracket
    Length (Metric) 7.62cm
    Reagent Type Halotron™ I
    UL Rating 2B:C
    Width (English) 6.38 in.
    Capacity (Metric) 1.1kg
    Product Type Fire extinguisher
    Height (English) 15.5 in.
    Height (Metric) 39.3cm
    Length (English) 3 in.
    Material Steel
    Model B385TS
    Range 6 to 8 ft.
    Temperature Range (English) -40° to 120°F
    Temperature Range (Metric) -40° to 48.8°C
    Width (Metric) 16.2cm
    Capacity (English) 2.5 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit