Amerex™ Halotron I Clean-Agent Stored-Pressure Fire Extinguishers

Most widely used Halon 1211 replacement agent

Manufacturer: Amerex CorporationSupplier Diversity Partner A384T

 View more versions of this product

Catalog No. 19-860-529

  Check Availability
Add to Cart

    Specifications

    Capacity 1.4 lb., 0.63kg
    Dimensions (L x W x H) 2.63 x 3.5 x 10 in. (6.6 x 8.8 x 25.4cm)
    Discharge Time 9 sec.
    Height (Metric) 25.4cm
    Reagent Type Halotron™ I
    UL Rating 1B:C
    Width (Metric) 8.8cm
    Capacity (Metric) 0.63kg
    Product Type Fire extinguisher
    Height (English) 10 in.
    Includes Anodized Aluminium valve, nozzle, vehicle bracket
    Length (English) 2.63 in.
    Length (Metric) 6.6cm
    Material Steel
    Model A384T
    Range 6 to 8 ft.
    Temperature Range (English) -40° to 120°F
    Temperature Range (Metric) -40° to 48.8°C
    Width (English) 3.5 in.
    Capacity (English) 1.4 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit