Amerex™ Halotron I Clean-Agent Stored-Pressure Fire Extinguishers

Most widely used Halon 1211 replacement agent

Manufacturer: Amerex CorporationSupplier Diversity Partner B394TS

 View more versions of this product

Catalog No. 19-860-530

  Check Availability
Add to Cart

    Specifications

    Capacity 5.5 lb., 2.49kg
    Dimensions (L x W x H) 4.25 x 5.13 x 15 in. (10.7 x 13 x 38cm)
    Reagent Type Halotron™ I
    Width (Metric) 13cm
    Product Type Fire extinguisher
    Discharge Time 9 sec.
    Height (English) 15 in.
    Height (Metric) 38cm
    Includes Anodized Aluminium valve, nozzle, aircraft bracket
    Length (English) 4.25 in.
    Length (Metric) 10.7cm
    Material Steel
    Model B394TS
    Range 9 to 15 ft.
    Temperature Range (English) -40° to 120°F
    Temperature Range (Metric) -40° to 48.8°C
    UL Rating 5B:C
    Width (English) 5.13 in.
    Capacity (English) 5.5 lb.
    Capacity (Metric) 2.49kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit