Amerex™ Halotron I Clean-Agent Stored-Pressure Fire Extinguishers Available on GSA/VA Contract for Federal Government customers only.

Most widely used Halon 1211 replacement agent

Manufacturer: Amerex CorporationSupplier Diversity Partner 397

 View more versions of this product

Catalog No. 19-805-993

  Check Availability
Add to Cart

    Specifications

    Height (English) 21.88 in.
    Length (English) 6 in.
    Reagent Type Halotron™ I
    UL Rating 1A:10B:C
    Capacity 11 lb., 4.9kg
    Dimensions (L x W x H) 6 x 9.25 x 21.88 in. (15.2 x 23.4 x 55.5cm)
    Discharge Time 9 sec.
    Height (Metric) 55.5cm
    Includes Anodized Aluminium valve, nozzle, wall bracket
    Length (Metric) 15.2cm
    Material Steel
    Model 397
    Temperature Range (English) -40° to 120°F
    Width (English) 9.25 in.
    Width (Metric) 23.4cm
    Capacity (Metric) 4.9kg
    Product Type Fire extinguisher
    Range 9 to 15 ft.
    Temperature Range (Metric) -40° to 48.8°C
    Capacity (English) 11 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit