Amerex™ Heavy-Duty Brackets for Fire Extinguishers Available on GSA/VA Contract for Federal Government customers only.

For use with a variety of Amerex fire extinguishers

Manufacturer: Amerex CorporationSupplier Diversity Partner 862

 View more versions of this product

Catalog No. 19-806-018

  Check Availability
Add to Cart

    Specifications

    Color Red
    Diameter (Metric) 12.7cm
    For Use With (Equipment) 13 lb. fire extinguisher
    Material Steel
    Product Type Heavy duty rubber strap bracket

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit