Learn More
Amerex™ Heavy-Duty Brackets for Fire Extinguishers
For use with a variety of Amerex fire extinguishers
Manufacturer:  Amerex™ 810
 View more versions of this product
Catalog No. 19-046986
Add to cart
Description
Description
  • Made of steel with a primed and powder-coated paint finish
  • Red brackets are galvanized
  • Designed to provide years of use
Compliance
Meet UL salt spray requirement (240 hr. @ 20% salt solution - ASTM B117)
Specifications
Specifications
Black
20 lb. fire extinguisher
Heavy duty metal strap bracket
17.7cm
Steel
Safety