Learn More
Amerex™ Heavy-Duty Brackets for Fire Extinguishers
For use with a variety of Amerex fire extinguishers
Manufacturer:  Amerex™ 5530
 View more versions of this product
Catalog No. 19-860-553
Add to cart
Description
Description
  • Made of steel with a primed and powder-coated paint finish
  • Red brackets are galvanized
  • Designed to provide years of use
Compliance
Meet UL salt spray requirement (240 hr. @ 20% salt solution - ASTM B117)
Specifications
Specifications
Heavy-duty Metal Strap Bracket
Steel
1 lb. fire extinguisher
Red
7.3 cm
845