Learn More
Amerex™ Heavy-Duty Brackets for Fire Extinguishers
For use with a variety of Amerex fire extinguishers
Supplier:  Amerex™ 0122P001
 View more versions of this product
Catalog No. 19-860-554
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Made of steel with a primed and powder-coated paint finish
  • Red brackets are galvanized
  • Designed to provide years of use
Compliance
Meet UL salt spray requirement (240 hr. @ 20% salt solution - ASTM B117)
Specifications
Specifications
Heavy-duty Metal Strap Bracket
Steel
5 lb. fire extinguisher
UL 299/UL 711
Red
10.7 cm
818