Amerex™ Purple K Dry Chemical Fire Extinguishers Available on GSA/VA Contract for Federal Government customers only.

Fire-fighting agent for the oil, gas, chemical and utilities industries

Manufacturer: Amerex CorporationSupplier Diversity Partner 497

 View more versions of this product

Catalog No. 19-035-398

  Check Availability
Add to Cart

    Specifications

    Capacity 50 lb., 22.6kg
    Diameter (English) Hose 0.5 in.
    Dimensions (L x W x H) 24 x 21.5 x 46 in. (61 x 55 x 117cm)
    Discharge Time 35 sec.
    Height (English) 46 in.
    Includes Brass valve and wall bracket
    Length (English) Hose 25 ft.
    Length (English) 24 in.
    Length (Metric) Hose 8m
    Length (Metric) 61cm
    Material Steel
    Model 497
    Reagent Type Purple K
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -53.8° to 48.8°C
    UL Rating 160B:C
    Width (English) 21.5 in.
    Width (Metric) 55cm
    Capacity (English) 50 lb.
    Capacity (Metric) 22.6kg
    Product Type Fire extinguisher
    Diameter (Metric) Hose 1.27cm
    Height (Metric) 117cm
    Range 25 to 35 ft.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit