Amerex™ Regular Dry Chemical Fire Extinguisher

Easy and economical to maintain and service

Manufacturer: Amerex CorporationSupplier Diversity Partner B403T

 View more versions of this product

Catalog No. 19-860-501

  Check Availability
Add to Cart

    Specifications

    Capacity 2.5 lb., 1.1kg
    Range 9 to 15 ft.
    Width (English) 5.25 in.
    Capacity (Metric) 1.1kg
    Dimensions (L x W x H) 4.25 x 5.25 x 15.25 in. (10.7 x 13.3 x 38.7cm)
    Discharge Time 10 sec.
    Height (English) 15.25 in.
    Height (Metric) 38.7cm
    Includes Wall bracket/vehicle
    Length (English) 4.25 in.
    Length (Metric) 10.7cm
    Material Steel
    Model B403T
    Reagent Type Regular dry chemical
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -53.8° to 48.8°C
    UL Rating 10B:C
    Width (Metric) 13.3cm
    Capacity (English) 2.5 lb.
    Product Type Fire extinguisher

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit