Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ Wet Chemical Fire Extinguishers

Supplier:  Amerex™ 16112

 View more versions of this product

Catalog No. 19-860-528


Add to Cart

Description

Description

Specifications

Specifications

Fire Extinguisher
2.5 gal.
7 x 9 x 24.5 in. (17.78 x 22.8 x 62.2 cm)
24.5 in.
Wall bracket
17.78 cm
10 to 12 ft.
40° to 120°F
2A:K
22.8 cm
Stainless Steel
9.4 L
107 sec.
62.2 cm
7 in.
B262
Potassium Acetate solution
4.4°C to 48.8°C
9 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit