Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amerex™ Wheeled CO2 Fire Extinguishers
For Class B and C fires
Supplier: Amerex™ 9331
Includes: Supplied with a 0.25 in. dia. (63mm) reinforced wire braided, shock-resistant hose with large brass handle and chrome-plated squeeze grip hose shut-off valve
Description
- Utilizes clean, odorless carbon dioxide gas to extiguish fires from flammable liquids and gases and electrical sources
- Features a durable brass, chrome-plated valve with easy operating “T” type operating handle
- Heavy-duty steel carriage with two easy-rolling, all-weather 4W x 16 in. dia. full-pneumatic rubber wheels
- Corrosion and impact-resistant red polyurethane enamel paint finish
Order Info
Discharge time: 44 sec. (50 lb.); 74 sec. (100 lb.); Discharge range: 10–15 ft. (3–4.5m); Operating temperature range: -40–+120°F ( -40–+49°C)
Certifications
U/L rating: 20 B:C
Compliance
Coast Guard approved.
Specifications
Wheeled Fire Extinguisher | |
0.25 in. | |
48 x 23 x 47 in. (122 x 58 x 120 cm) | |
47 in. | |
0.25 in. dia. reinforced wire braided, shock resistant hose with large Brass handle and Chrome plated squeeze grip hose shut-off valve | |
15 ft. | |
4.5 m | |
10 to 15 ft. | |
-40° to +120°F | |
20B:C | |
45.3 kg | |
58 cm |
Steel | |
0.6 cm | |
74 sec. | |
120 cm | |
48 in. | |
122 cm | |
334 | |
Carbon Dioxide | |
-40°C to +48.8°C | |
100 lb. | |
23 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title