Amerex™ Wheeled Fire Extinguishers: Multipurpose ABC Dry Chemical

Wheeled, dry chemical fire extinguisher

Manufacturer: Amerex CorporationSupplier Diversity Partner 467

 View more versions of this product

Catalog No. 19-860-544

  Check Availability
Add to Cart

    Specifications

    Capacity 125 lb., 57kg
    Diameter (English) Hose 0.75 in.
    Dimensions (L x W x H) 38 x 26 x 52 in. (96.5 x 66 x 132cm)
    Discharge Time 48 sec.
    Length (English) Hose 50 ft.
    Length (English) 38 in.
    UL Rating 40A:240B:C
    Width (English) 26 in.
    Capacity (Metric) 57kg
    Diameter (Metric) Hose 1.9cm
    Height (English) 52 in.
    Height (Metric) 132cm
    Length (Metric) Hose 15m
    Length (Metric) 96.5cm
    Model 467
    Range 30 to 40 ft.
    Reagent Type ABC dry chemical
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -54° to 49°C
    Weight (English) 404 lb.
    Weight (Metric) 183kg
    Width (Metric) 66cm
    Capacity (English) 125 lb.
    Product Type Semi pneumatic wheel fire extinguisher

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit