Learn More
Amerex™ Wheeled Fire Extinguishers: Multipurpose ABC Dry Chemical
Wheeled, dry chemical fire extinguisher
Supplier: Amerex™ 10701
Description
- Semi-pneumatic wheels
- Coast Guard approved
- Contains a specially fluidized and siliconized mono ammonium phosphate chemical
- Chemically insulates Class A fires by melting at approximately 350°F, coating the surface to which it is applied
- Smothers and breaks the chain reaction of Class B fires and will not conduct electricity
Specifications
Steel-wheeled Fire Extinguisher | |
57 kg | |
1.9 cm | |
48 sec. | |
132 cm | |
50 ft. | |
15 m | |
30 to 40 ft. | |
-65° to +120°F | |
40A:240B:C | |
196 kg | |
71 cm |
125 lb. | |
0.75 in. | |
45 x 28 x 52 in. (114 x 71 x 132 cm) | |
52 in. | |
45 in. | |
114 cm | |
470 | |
ABC dry chemical | |
-54°C to +49°C | |
432 lb. | |
28 in. |