Learn More
Amerex™ Wheeled Fire Extinguishers: Multipurpose ABC Dry Chemical
Wheeled, dry chemical fire extinguisher
Manufacturer:  Amerex™ 11677
 View more versions of this product
Catalog No. 19-860-550
Add to cart
Description
Description
  • Semi-pneumatic wheels
  • Coast Guard approved
  • Contains a specially fluidized and siliconized mono ammonium phosphate chemical
  • Chemically insulates Class A fires by melting at approximately 350°F, coating the surface to which it is applied
  • Smothers and breaks the chain reaction of Class B fires and will not conduct electricity
Specifications
Specifications
Steel-wheeled Fire Extinguisher
136 kg
2.54 cm
60 sec.
145 cm
50 ft.
15 m
30 to 40 ft.
-65° to +120°F
40A:320B:C
337 kg
88 cm
300 lb.
1 in.
39 x 34 x 57 in. (99 x 88 x 145 cm)
57 in.
39 in.
99 cm
491
ABC dry chemical
-54°C to +49°C
743 lb.
34 in.