Amerex™ Wheeled Fire Extinguishers: Multipurpose ABC Dry Chemical

Wheeled, dry chemical fire extinguisher

Manufacturer: Amerex CorporationSupplier Diversity Partner 450

 View more versions of this product

Catalog No. 19-860-542

  Check Availability
Add to Cart

    Specifications

    Capacity 125 lb., 57kg
    Diameter (Metric) Hose 1.9cm
    Dimensions (L x W x H) 36 x 24 x 45 in. (91.4 x 61 x 114.3cm)
    Length (English) 36 in.
    Range 30 to 40 ft.
    Weight (Metric) 152kg
    Capacity (Metric) 57kg
    Diameter (English) Hose 0.75 in.
    Discharge Time 52 sec.
    Height (English) 45 in.
    Height (Metric) 114.3cm
    Length (English) Hose 50 ft.
    Length (Metric) Hose 15m
    Length (Metric) 91.4cm
    Model 450
    Reagent Type ABC dry chemical
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -54° to 49°C
    UL Rating 40A:240B:C
    Weight (English) 335 lb.
    Width (English) 24 in.
    Width (Metric) 61cm
    Capacity (English) 125 lb.
    Product Type Semi pneumatic wheel fire extinguisher

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit