Amerex™ Wheeled Fire Extinguishers: Purple K

Wheeled, dry chemical fire extinguisher

Manufacturer: Amerex CorporationSupplier Diversity Partner 490

 View more versions of this product

Catalog No. 19-860-541

  Check Availability
Add to Cart

    Specifications

    Capacity 125 lb., 57kg
    Dimensions (L x W x H) 40 x 28 x 53.5 in. (101.6 x 71 x 135.8cm)
    Discharge Time 48 sec.
    Length (Metric) Hose 15m
    Length (Metric) 101.6cm
    Reagent Type ABC dry chemical
    Temperature Range (English) -65° to 120°F
    UL Rating 320B:C
    Weight (English) 344 lb.
    Weight (Metric) 156kg
    Width (English) 28 in.
    Capacity (English) 125 lb.
    Capacity (Metric) 57kg
    Diameter (English) Hose 0.75 in.
    Height (English) 53.5 in.
    Height (Metric) 135.8cm
    Length (English) Hose 50 ft.
    Length (English) 40 in.
    Range 30 to 40 ft.
    Temperature Range (Metric) -54° to 49°C
    Product Type Semi pneumatic wheel fire extinguisher
    Diameter (Metric) Hose 1.9cm
    Model 490
    Width (Metric) 71cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit