Learn More
Amerex™ Wheeled Fire Extinguishers: Purple K
Wheeled, dry chemical fire extinguisher
Manufacturer:  Amerex CorporationSupplier Diversity Partner 493
 View more versions of this product
Catalog No. 19-860-552
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Steel Wheeled fire extinguisher
1 in.
39 x 34 x 57 in. (99 x 88 x 145cm)
57 in.
50 ft.
15m
493
Purple K
-54° to 49°C
743 lb.
34 in.
300 lb.
300 lb., 136kg
2.54cm
67 sec.
145cm
39 in.
99cm
30 to 40 ft.
-65° to 120°F
320B:C
337kg
88cm
136kg