Learn More
Amerex™ Wheeled Fire Extinguishers: Purple K
Wheeled, dry chemical fire extinguisher
Supplier: Amerex™ 15502
Description
- Purple-K chemical extinguisher
- Semi-pneumatic wheels
- Coast Guard approved
Specifications
Semi-pneumatic Wheeled Fire Extinguisher | |
1.9 cm | |
52 sec. | |
132 cm | |
50 ft. | |
15 m | |
30 to 40 ft. | |
-65° to +120°F | |
320B:C | |
57 kg | |
66 cm |
0.75 in. | |
38 x 26 x 52 in. (96.5 x 66 x 132 cm) | |
52 in. | |
38 in. | |
96.5 cm | |
469 | |
Purple K | |
-54°C to +49°C | |
125 lb. | |
26 in. |