Learn More
Amerex™ Wheeled Fire Extinguishers: Purple K
Wheeled, dry chemical fire extinguisher
Supplier:  Amerex™ 17483
 View more versions of this product
Catalog No. 19-860-549
Add to Cart
Description
Description
  • Purple-K chemical extinguisher
  • Semi-pneumatic wheels
  • Coast Guard approved
Specifications
Specifications
Steel-wheeled Fire Extinguisher
1.9 cm
52 sec.
132 cm
50 ft.
15 m
30 to 40 ft.
-65° to +120°F
320B:C
57 kg
71 cm
0.75 in.
45 x 28 x 52 in. (114 x 71 x 132 cm)
52 in.
45 in.
114 cm
472
Purple K
-54°C to +49°C
125 lb.
28 in.