Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amerex™ Wheeled Fire Extinguishers: Purple K
Wheeled, dry chemical fire extinguisher
$7930.00 - $14250.00
Specifications
Length (English) Hose | 50 ft. |
---|---|
Length (Metric) Hose | 15m |
Range | 30 to 40 ft. |
Temperature Range (English) | -65° to 120°F |
Temperature Range (Metric) | -54° to 49°C |
Catalog Number | Mfr. No. | Capacity (English) | Product Type | Discharge Time | Reagent Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Product Type | Discharge Time | Reagent Type | Price | Quantity & Availability | ||
19-065-597
|
Amerex™
10643 |
125 lb. | Semi pneumatic wheel fire extinguisher | 53 sec. | Purple K |
Each for $8,680.00
|
|
19-860-541
|
Amerex™
10490 |
125 lb. | Semi pneumatic wheel fire extinguisher | 48 sec. | ABC dry chemical |
Each for $7,930.00
|
|
19-860-546
|
Amerex™
15502 |
125 lb. | Semi pneumatic wheel fire extinguisher | 52 sec. | Purple K |
Each for $9,960.00
|
|
19-860-549
|
Amerex™
17483 |
125 lb. | Steel Wheeled fire extinguisher | 52 sec. | Purple K |
Each for $12,930.00
|
|
19-860-552
|
Amerex™
15505 |
300 lb. | Steel Wheeled fire extinguisher | 67 sec. | Purple K |
Each for $14,250.00
|
|
Description
- Purple-K chemical extinguisher
- Semi-pneumatic wheels
- Coast Guard approved
Specifications
50 ft. | |
30 to 40 ft. | |
-54° to 49°C |
15m | |
-65° to 120°F | |
320B:C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title