Amerex™ Wheeled Fire Extinguishers: Regular Dry Chemical

Wheeled, dry chemical fire extinguisher

Manufacturer: Amerex CorporationSupplier Diversity Partner 451

 View more versions of this product

Catalog No. 19-860-543

  Check Availability
Add to Cart

    Specifications

    Capacity 150 lb., 68kg
    Diameter (English) Hose 0.75 in.
    Diameter (Metric) Hose 1.9cm
    Dimensions (L x W x H) 36 x 24 x 45 in. (91.4 x 61 x 114.3cm)
    Discharge Time 60 sec.
    Height (English) 45 in.
    Height (Metric) 114.3cm
    Length (English) Hose 50 ft.
    Length (English) 36 in.
    Length (Metric) Hose 15m
    Length (Metric) 91.4cm
    Range 30 to 40 ft.
    Reagent Type Regular dry chemical
    Temperature Range (English) -65° to 120°F
    Temperature Range (Metric) -54° to 49°C
    UL Rating 240B:C
    Weight (English) 360 lb.
    Weight (Metric) 163kg
    Width (English) 24 in.
    Width (Metric) 61cm
    Capacity (Metric) 68kg
    Product Type Semi pneumatic wheel fire extinguisher
    Model 451
    Capacity (English) 150 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit