Amerex™ Wheeled Fire Extinguishers: Regular Dry Chemical Available on GSA/VA Contract for Federal Government customers only.

Wheeled, dry chemical fire extinguisher

Manufacturer: Amerex CorporationSupplier Diversity Partner 496

 View more versions of this product

Catalog No. 19-035-397

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Hose 0.5 in.
    Capacity (English) 50 lb.
    Diameter (Metric) Hose 1.27cm
    Capacity (Metric) 23kg
    Discharge Time 35 sec.
    Height (English) 46 in.
    Height (Metric) 117cm
    Length (Metric) 61cm
    Length (English) 24 in.
    Length (Metric) Hose 8m
    Length (English) Hose 25 ft.
    Product Type Semi pneumatic wheel fire extinguisher
    Model 496
    Temperature Range (English) -40° to 120°F
    Range 25 to 35 ft.
    Reagent Type Regular dry chemical
    UL Rating 160B:C
    Weight (English) 174 lb.
    Weight (Metric) 79kg
    Width (Metric) 55cm
    Width (English) 21.5 in.
    Temperature Range (Metric) -40° to 49°C
    Capacity 50 lb., 23kg
    Dimensions (L x W x H) 24 x 21.5 x 46 in. (61 x 55 x 117cm)

    • Semi-pneumatic wheels
    • Coast Guard approved
    • Contains a siliconized sodium bicarbonate-based dry chemical with free flowing and non-caking additives
    • Economical Class B and C protection with lower initial cost and recharging
    • Chemical smothers fires in flammable liquids and pressurized gases and is electrically nonconductive

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit