Learn More
American Educational Products Dissection Trays
Ideal for general student use.
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
S65104
![]() |
American Educational Products
7355 |
Deluxe Tray |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
Durable, one-piece aluminum dissection trays won't rust, crack or leak.
- Standard-size tray measures 11.5 x 7.5 in. (28.8 x 18.9cm)
- Deluxe tray measures 13.13 x 9.38 x 2.25 in. (33 x 23.5 x 5.6cm)
- Both come lined with black wax that can be reused or resurfaced for continual use
Specifications
Aluminum |