Learn More
American Educational Products Low-Form Crucibles Products for Science Education
Highly resistant to alkalis and hot temperatures.
Specifications
Finish Glazed
Material Porcelain
Description With Lid cover
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
S326461 Products for Science Education
View Documents
American Educational Products
7S326461
30 mL 51 mm 30 mm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Glazed inside and out except for outside bottom surface
  • With cover
  • Pack of 10
Specifications
Specifications
Glazed
Porcelain
With Lid cover