Learn More
American Educational Products Lung Demonstration Apparatus
Relaxation and contraction illustrated.
Catalog Number | Mfr. No. | Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Application) | Price | Quantity & Availability | ||||
S41390
![]() |
American Educational Products
712033 |
Lung replacement Rubber dam | Simple model demonstrates the mechanisms of breathing for students |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
This simple model demonstrates the mechanisms of breathing for students.
- Shows how the lungs are emptied and filled with air by the relaxation and contraction of the diaphragm muscle
- One or two balloons attached to a Y-tube simulate the lungs
- The chest cavity is a half-gallon bell jar with open top and stout rubber sheet that acts as the diaphragm
- As the diaphragm is pulled downward, the atmospheric pressure is lowered inside the chamber, causing the lungs to expand and take in air
- When the diaphragm is released, the lungs deflate
Specifications
Plastic |