Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Solid Rubber Stoppers
SCI_ED

Pliable, noncorrosive and deformation resistant.

Specifications

Color Black
Material Rubber
Product Type Specs Solid Stopper
Stopper Number 0
For Use With (Application) Pliable, noncorrosive and deformation resistant
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Bottom Diameter (Metric) Inner Quantity Stopper Number Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Bottom Diameter (Metric) Inner Quantity Stopper Number Price Quantity & Availability  
S50080A
SCI_ED
View Documents
American Educational Products
7S50080A
1.2 cm 1.5 cm 105 0 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
S50080B
SCI_ED
View Documents
American Educational Products
7S50080B
1.6 cm 1.8 cm 77 0 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

 

  • Number per pound (454g) is based on solid stoppers
  • Color: Black

 

Specifications

Specifications

Black
Solid Stopper
Pliable, noncorrosive and deformation resistant
Rubber
0
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit