Learn More
American Educational Products Strong Ring Supports Products for Science Education
Cast-iron construction prevents breakage under normal use.
Specifications
Finish Black Enamel
For Use With (Application) Prevents breakage under normal use
Includes Clamp
Material Cast Iron
Packaging Quantity Each
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
S47791 Products for Science Education
View Documents
American Educational Products
7S47791
4.4 in. 5 in. 11.1cm 12.7cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Coated with black enamel that withstands chemical deterioration
  • Sturdy thumbscrew in an integral clamp locks the ring tightly to support stands, lattices, etc.
  • Ring center is a standard 4.2 in. (10.8cm) from support rod center
  • S47788 ring has lugs for holding crucibles
Specifications
Specifications
Black Enamel
Clamp
Each
Prevents breakage under normal use
Cast Iron
Support Stand