Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Two-Hole Rubber Stoppers Products for Science Education

Pliable, noncorrosive and deformation resistant.

Manufacturer:  American Educational Products 7S50082F

 View more versions of this product

Catalog No. S50082F



Add to cart

Description

Description

  • Each stopper displays its size number in raised figures on top
  • Color: Black

Specifications

Specifications

0.5 cm
Pliable, noncorrosive and deformation resistant
33
2
Rubber
2.6 cm
For 2 to 2.3cm I.D. openings
4
Black
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit