Learn More
AccuTec Blades American Line™ Heavy Duty Snap-Off Knife
AccuTec Blades American Line™ Heavy Duty Snap-Off Knife is ergonomically designed to fit in the palm for excellent cutting control
$5.93 - $8.84
Specifications
For Use With (Application) Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Quantity 1 Knife, 3 Blades
Material Carbon Steel
Sterility Non-sterile
Type Heavy-Duty Knife
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
17-989-135
View Documents
AccuTec Blades
660397
8 Point / 18 mm
Each for $8.84
Pack of 5 EA for $35.35
Add to cart
17-989-134
View Documents
AccuTec Blades
660399/BADUOM
13 Point / 9 mm
Each for $5.93
Pack of 5 EA for $23.70
Add to cart
Description
Description
  • Includes 1 Heavy Duty Knife, and 3 Blades
  • Metal track and body
  • Textured slide for easy blade extension and retraction
  • Ergonomic rubber like handle for comfortable grip
  • Autolock slider locks blade instantly for safety and convenience
Specifications
Specifications
Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Carbon Steel
Heavy-Duty Knife
1 Knife, 3 Blades
Non-sterile