American Lock™ Lockout Hasp

Manufacturer: American Lock Company LO80

 View more versions of this product

Catalog No. 19-041-621

  Check Availability
Add to Cart

    Specifications

    Holds 6 locks
    Product Type Lockout Hasps
    Color Red
    Diameter (English) 1 in.
    Diameter (Metric) 3cm
    For Use With (Application) Locking tabs for extra pry resistance
    Material Aluminum

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit