Learn More
GE Healthcare Amersham CyDye bis-Reactive Dye Available on GSA/VA Contract for Federal Government customers only.
Designed for a convenient way to label with CyDye Fluorescent Dyes. GE Healthcare Amersham CyDye bis-Reactive Dye is packaged in premixed amounts and foil-sealed to ensure consistent labeling. Each kit contains five vials of dye.
Manufacturer:  GE Healthcare PA25500
Catalog No. 45-200-575
Add to cart
Description
Description
Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes
  • Ideal for use in ELISA, immunoprecipitation and blotting applications
  • Contains fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups
  • Kit contains five vials of dye: each vial is sufficient for labeling 1mg of protein or 30nmol of amino groups
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Five foil packs each containing dried dye to label 1 mg of protein.
For the labelling of biological compounds with Cy5.5 bisfunctional dye
Cy5.5 bis-Reactive Dye Pack
1 Kit
Blue
Cy5.5 bisfunctional dye
2 - 8°C, protected from light
1311.58