Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Amersham CyDye DIGE Fluor Dye, Cy2 Available on GSA/VA Contract for Federal Government customers only.
Designed for use in 2-D Fluorescence Difference Gel Electrophoresis using the Ettan DIGE system. Cytiva Amersham CyDye DIGE Fluor minimal dyes and saturation dyes are size- and charge-matched fluorescent dyes specifically engineered for detecting protein abundance differences.
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 25800860
Catalog No. 45-002-403
Add to cart
Description
Description
CyDye DIGE fluors are exceptional dyes for multicolor analysis, offering bright and intense colors with narrow excitation and emission bands. The fluors are spectrally distinct, making them highly suitable for multicolor detection. CyDye DIGE fluors utilize these benefits, but are also size- and charge-matched specifically for 2-D DIGE using the Ettan DIGE system.
  • Allows detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
Available as minimal and saturation labeling dyes
  • The minimal dyes are intended for general 2-D application use where sufficient amounts of sample are available
  • The saturation dyes from the Scarce Sample Labeling Kit are designed to be used for applications where only small amounts of sample are available, for example in Laser Capture Microdissection
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yellow
10 nmol (2 x 5 nmol) of CyDye DIGE fluor Cy2 minimal dye
10 nmol
Cy2
−15° to −30°C , Avoid light, store in the dark.