Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

GE Healthcare Amersham CyDye Fluorescent Nucleotides Available on GSA/VA Contract for Federal Government customers only.

Ideal for multiplexing within a single sample. GE Healthcare Amersham CyDye Fluorescent Nucleotides are fluorescent dyes with superior characteristics for genomic and protein research. Available in multiple sizes and colors, including a multipack option for maximum versatility.

Manufacturer:  GE Healthcare PA55032

 View more versions of this product

Catalog No. 45-200-577



Description

Description

CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. They are ideal for labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands, well suited for multiplexing within a single sample. They exhibit the following properties:
  • High sensitivity
  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Fluorescence Color: Far-red
250 nmol
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit