Learn More
GE Healthcare Amersham CyDye Value Packs (bis-Reactive NHS Ester), Blue Available on GSA/VA Contract for Federal Government customers only.
Label proteins, antibodies and more. GE Healthcare Amersham CyDye Value Packs (bis-Reactive NHS Ester) provide maximum economy in general labeling applications. Available in a variety of reactive dye colors: red (Cy3), blue (Cy5.5) and purple (Cy7).
Manufacturer:  GE Healthcare PA15504
Catalog No. 45-002-449
Add to cart
Description
Description
Fluorescent dyes are a convenient way to label antibodies, proteins, drugs, and oligonucleotides. They are ideal for use in ELISA, immunoprecipitation and blotting applications.
  • Suitable for general labeling of protein amine groups
  • Used for high-throughput screening (HTS) applications
  • Bulk packs for greatest economy
  • Foil packs of fluors carrying two reactive groups on each dye molecule
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Foil packs, containing dried dye.
Used for high-throughput screening (HTS) applications
Cy5.5 Bis NHS Ester
10 mg
Blue
Cy5.5
2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
1311.58