Learn More
GE Healthcare Amersham CyDye Value Packs (Mono-Reactive Maleimide), Cy3 Available on GSA/VA Contract for Federal Government customers only.
Designed for selective labeling of thiol groups, including free sulfhydryl groups and antibodies. GE Healthcare Amersham CyDye Value Packs (Mono-Reactive Maleimide) provide maximum economy, convenience and quality.
Manufacturer:  GE Healthcare PA13130
Catalog No. 45-002-448
Add to cart
Description
Description
CyDye fluors are bright fluorescent dyes with good aqueous solubility, pH insensitivity, low non-specific binding, and photostability. They are an excellent choice for use in steady state fluorescence, FP and FRET applications. Mono-reactive maleimides enable labeling via thiol groups, when labelling via amine groups is not possible or desirable.
  • Ideal for selective labeling of free sulfhydryl groups, such as cysteine residues in peptides, proteins, and oligonucleotides
  • Particularly suited for antibody labeling, since the dye is introduced away from the antibody binding site, decreasing the chance that the functionality of the labeled molecule will be impaired
  • Bulk packs provide the greatest economy
  • Example protocols are included with each dye pack
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Foil pack containing 5mg dried dye.
For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications
Cy3 Maleimide
5mg
Soluble in cold water and hot water
Red
Cy3
2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
765.95