Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
GE Healthcare Amersham™ ECL™ DualVue Western Blotting Markers PROMO
Detect proteins on gel and membrane as well as on film
Manufacturer:  GE Healthcare 29334869
Catalog No. 45-001-182PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Confirm protein transfer, definite blot orientation and permit calculation of target protein's molecular mass.
  • Prestained markers indicate efficiency of protein transfer to membrane and blot orientation
  • Tagged recombinant protein markers allow accurate calculation of molecular weight of target proteins
  • Compatible with any HRP substrate as well as both PVDF and nitrocellulose membrane types
  • Contain a mixture of three prestained colored markers of defined molecular weight (from 15,000 to 100,000) and seven tagged recombinant proteins (molecular weights from 15,000 to 150,000)
Specifications
Specifications
25 Loadings
Liquid
Protein Molecular Weight Marker/Ladder
Detect proteins on Gel and membrane as well as on film
−15° to −30°C
Safety and Handling
Safety and Handling
ShelfLife : 3 Months