Learn More
GE Healthcare Amersham™ ECL™ DualVue Western Blotting Markers: PROMO
Detect proteins on gel and membrane as well as on film
Manufacturer:  GE Healthcare RPN810PM
Catalog No. 45-001-182LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Confirm protein transfer, definite blot orientation and permit calculation of target protein's molecular mass.
  • Prestained markers indicate efficiency of protein transfer to membrane and blot orientation
  • Tagged recombinant protein markers allow accurate calculation of molecular weight of target proteins
  • Compatible with any HRP substrate as well as both PVDF and nitrocellulose membrane types
  • Contain a mixture of three prestained colored markers of defined molecular weight (from 15,000 to 100,000) and seven tagged recombinant proteins (molecular weights from 15,000 to 150,000)
Order Info
Each pack contains ECL DualVue markers (125μL) and S-protein-HRP conjugate (50μL) sufficient for 25 standard gel loadings.
Specifications
Specifications
ECL DualVue Markers: 125 μL, S-Protein-HRP Conjugate: 50 μL, sufficient for 25 standard gel loadings
Liquid
−15° to −30°C
Detect proteins on Gel and membrane as well as on film
Pre-Stained Indicator Proteins, Phosphorylase b: 50μg, 100000 (Mr), Rabbit muscle (source), Myoglobin: 50μg, 16000 (Mr), Horse muscle (source), Lysozyme: 50μg, 15000 (Mr), Chicken Egg white (source)
Protein Molecular Weight Marker/Ladder
Safety and Handling
Safety and Handling
ShelfLife : 3 Months