Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Amersham™ ECL™ Plex Fluorescent Rainbow Markers: PROMO

Product

Manufacturer:  GE Healthcare RPN850EPM

Catalog No. 45-001-594LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining
  • Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels
  • Ready-to-load
  • Provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination
  • Provides fluorescent images using Cy3 and Cy5 channels

Order Info

Each 120μL pack size is sufficient for use with at least 40 minigels (10 × 8cm) or 20 large gels (20 × 20cm). Each 500μL pack size is sufficient for use with at least 160 minigels (10 x 8cm) or 80 large gels (20 x 20cm).

Specifications

Specifications

120 μL
Quantitative western blotting
Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent
12 Minigel Loadings
Multicolor
Liquid
−20°C
Protein Molecular Weight Marker/Ladder
Safety and Handling

Safety and Handling

ShelfLife : 3 Months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit