Learn More
GE Healthcare Amersham™ ECL™ start Western Blotting Detection Reagent
Use this luminol-based chemiluminescent substrate for the detection of proteins immobilized to a membrane. GE Healthcare Amersham™ ECL™ start Western Blotting Detection Reagent has a long signal duration, which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.
Manufacturer:  GE Healthcare 29334868
Catalog No. 45-100-101PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Amersham™ ECL™ start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins.
  • Long signal duration provides assay design flexibility and robust detection of large experimental sets
  • Produces low background, regardless of method of detection
  • Working solution is stable for five days
  • Long shelf life of 18 hours is convenient for those who run Western blots infrequently
  • Detection on X-ray film or CCD Imager
  • Recommended Membrane: PVDF or nitrocellulose
  • For detection of high abundant proteins
  • Colorless detection reagent 1 and 2
Applications:
Routine and confirmatory Western blotting
Specifications
Specifications
Amersham™ ECL™ start Western Blotting Detection Reagent
400 mL
Western Blotting
4°C to 8°C