Learn More
GE Healthcare Amersham™ ECL™ Prime Western Blotting Detection Reagent: PROMO
Enable the use of highly diluted primary and secondary antibodies without reducing sensitivity with this detection reagent. GE Healthcare Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.
Manufacturer:  GE Healthcare RPN2236PM
Catalog No. 45-010-090LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: Solution A (luminol solution) and Solution B (peroxide solution)
Description
Description
  • Stable signal enables multiple exposures and makes reagent suitable for large experimental series
  • Provides convenient handling time between detection and completion of experiment
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
  • Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers
Education and research
Specifications
Specifications
Sensitive quantitative or confirmatory western blotting
Amersham ECL Prime Western Blotting Detection Reagent
3000cm2 membrane
Solution A (Luminol solution, 3 × 50mL) and Solution B (Peroxide solution, 3 × 50mL)
≈1 pg
2°C to 8°C